Learn More
Amsbio LLC (formerly Genlantis) EZ-Spread™ Plating Beads
Description
- Sterile plating in seconds
- No need for glass rods and potentially hazardous ethanol flaming
- More uniform spreading and more colonies compared to traditional glass rod method
- Two choices of ready-to-use sterile packaging: dispenser bottle for spreading 100 plates or single-use tubes for 50 plating
- Fast, easy way to spread cells on multiple plates simultaneously
- Safely spread bacterial and yeast cells across the surface of an agar plate
Specifications
Specifications
| For Use With (Application) | Easily and safely spread bacterial and yeast cells across the surface of an agar plate |
| Quantity | Each |