Learn More
Amsbio LLC (formerly Genlantis) EZ-Spread™ Plating Beads
Catalog No. C400100
Encompass
Change view
Click to view available options
Quantity:
50 Tubes
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C400100 Each
C400050 50 Tubes
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C400100 Supplier Amsbio LLC (formerly Genlantis) Supplier No. C400100
Only null left
Allows you to easily and safely spread bacterial and yeast cells across the surface of an agar plate
  • Sterile plating in seconds
  • No need for glass rods and potentially hazardous ethanol flaming
  • More uniform spreading and more colonies compared to traditional glass rod method
  • Two choices of ready-to-use sterile packaging: dispenser bottle for spreading 100 plates or single-use tubes for 50 plating
  • Fast, easy way to spread cells on multiple plates simultaneously
  • Safely spread bacterial and yeast cells across the surface of an agar plate
Specifications
For Use With (Application) Easily and safely spread bacterial and yeast cells across the surface of an agar plate
Quantity Each