Learn More
Genlantis™ Beta-Gal Assay Kits DFS Item
Sensitive and accurate quantitation of β-Gal expression
Supplier:  Genlantis™ A10100K
Catalog No. A10100K
Only null left
Explore available promotions
Description
Description
  • Quantitative, sensitive, and convenient assays for determining βgalactosidase expression
  • Complete kits with ready-to-use reagents, including the E. coli βgalactosidase enzyme standard
  • Suitable for direct transfection assays in 96-well plates
  • Elimination of freeze-thaw cycles
  • The βGalactosidase Assay Kits quantitatively measure the expression level of βgalactosidase in cells transfected with vectors carrying the lacZ gene
Specifications
Specifications
beta-Gal assay kit
Sensitive and accurate quantitation of beta-gal expression
500 Assays
500 Microassays in 96-well plate
100 μL beta-gal enzyme standard, 40U, 5 mL lysis buffer 5X, 5 x 1 mL 10X CPRG substrate stock, 55 mL standard dilution buffer, 55 mL stop buffer, 55 mL substrate buffer
-20°C
Product Suggestions
Product Suggestions