missing translation for 'onlineSavingsMsg'
Learn More

Genistein, ≥97% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111014217
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014217 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111014217 Supplier Merck Emd Millipore Supplier No. 9213610MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Genistein, a naturally occurring soy isoflavone, is a dietary protein tyrosine kinase (PTK) inhibitor. It is found to possess anticarcinogenic properties.

Featured Industries: Food and Beverages

Specifications

CAS 446-72-0
Physical Form Neat
Linear Formula C15H10O5
MDL Number MFCD00016952
Synonym 4′,5,7-Trihydroxyisoflavone; 5,7-Dihydroxy-3-(4-hydroxyphenyl)-4 H-1-benzopyran-4-one
Type Analytical Standard
Quantity 10 mg
Formula Weight 270.24
Percent Purity ≥97% (HPLC)
Grade Analytical Standard