missing translation for 'onlineSavingsMsg'
Learn More

Ion Torrent™ Genexus™ Templating Strips 3B-GX5 and 4

Catalog No. A49782
Change view
Click to view available options
Quantity:
2 runs kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A49782 2 runs kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A49782 Supplier Ion Torrent™ Supplier No. A49782

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Kit provides eight each of the strips required to perform automated template preparation and chip loading of two Ion Torrent™ GX5 chips using the Ion Torrent™ Genexus™ Integrated Sequencer. This kit is specifically for use with the Oncomine™ Myeloid GX V2 assay.

  • Kit provides eight each of the strips required to perform automated template preparation and chip loading of two GX5 chips using the Genexus Integrated Sequencer
  • This kit, combined with the GX5 Chip and Genexus Coupler (A40269) supports flexible, fully automated template preparation and chip loading of libraries prepared using the Genexus Integrated Sequencer or with an alternative protocol
  • Maximizes flexibility with the ability to load one to four chip lanes per run
  • Designed to enable cost-effective processing of 1 to 32 single pool libraries
  • Specifically for use with the Oncomine Myeloid GX v2 assay
  • Delivers fast and simple instrument setup, with less than five minutes of hands-on time
  • For all other Genexus assays, please use the Genexus Templating Strips 3-GX5 and 4 kit (A40263)
  • For research use only, not for use in diagnostic procedures

Specifications

Sample Type DNA and RNA from Blood or Bone Marrow
For Use With (Equipment) Genexus™ Integrated Sequencer
For Use With (Application) Sequencing
No. of Reactions 2 Runs
Quantity 2 runs kit