missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ion Torrent™ Genexus™ FFPE DNA and RNA Purification Kit
Description
- A set of consumables that enables sequential, automated extraction and quantitation of DNA and RNA from formalin-fixed, paraffin-embedded (FFPE) lysate from tumor resections, fine needle aspirates (FNA), and core needle biopsies (CNB)
- Kit supports four runs of up to 12 samples each, utilizing the proven MagMAX technology for extraction and Qubit technology for post-extraction nucleic acid quantitation
- Consumables are prefilled with most reagents necessary to perform the extraction, so only a few pipetting steps are required per sample to set up the run
- Simply remove the foil seal, add enzymes and samples, then place the consumables onto the instrument
- Achieve consistent extraction results with MagMAX technology
- Reduce hands-on setup time using pre-filled consumables
- Maximize efficiency with automated Qubit quantitation
- Experience rapid turnaround times of as little as two hours from lysate to quantified nucleic acid
- For research use only, not for use in diagnostic procedures
Spécifications
Spécifications
| Format | Pre-filled Reagent Plates |
| Type d’échantillon | Core Needle Biopsies, Fine Needle Aspirates (FNA), FFPE |
| À utiliser avec (équipement) | Genexus™ Purification System |
| À utiliser avec (application) | Next Generation Sequencing |
| Nb de réactions | 48 |
| Sortie | Purified, quantified DNA and/or RNA |
| Méthode de purification | MagMax™ bead-based purification |
| Shipping condition (Condition d'expédition) | Ambient |
| Étape du flux de travail | Sample Purification and Quantification |
For Research Use Only. Not for use in Diagnostic Procedures.