Learn More
Applied Biosystems™ Genetic Analyzer Upgrade Kit (3500 to 3500xL)
Description
This kit allows you to upgrade your 8 capillary 3500 Genetic Analyzer to a 24 capillary 3500xL expanding the capabilities of your instrument. You'll be able to take advantage of a dramatic increase in throughput and in overall laboratory productivity. The upgrade is simple and completed within a day. It does not involve any changes to the computer or software so you will not need to re-verify these components of your system.
- 8 capillary system can easily be upgraded to a 24-capillary system
- New single-line 505 nm, solid-state long-life laser—utilizes a standard power supply; requires no heat-removal ducting
- Powerful, integrated data collection and primary analysis software provides real-time assessment of data quality
- Radio Frequency Identification (RFID) technology tracks key consumables data and records administrative information
- Advanced multiplexing capabilities for DNA fragment analysis with up to six unique dyes
- Unrivaled application flexibility—one array and one polymer are used for most applications
- Simple setup, operation and maintenance—the easiest-to-run, easiest-to-own DNA sequencer to date
Specifications
Specifications
| Content And Storage | This upgrade comes with a starter kit. |
| Format | Kit |
| For Use With (Equipment) | 3500 Genetic Analyzer |
| For Use With (Application) | Sequencing |
| Product Type | Genetic Analyzer Upgrade Kit |
| Quantity | 1 Kit |
| System Upgrade | 3500 to 3500XL |
Frequently Asked Questions (FAQs)
If you are using the most recent version of Data Collection Software (v3.3), there is a way to automatically upload your data into the Thermo Fisher Cloud app from the instrument. When a run is complete, the data is stored in the results group folder that is specified in the plate record. The data will be automatically uploaded to your account if your User ID is the active ID. Results are saved in your account folders named by the plate and injections, not in the results group folder. Please go to page 43 of the 3500/3500xL Genetic Analyzer User Guide for Data Collection Software v3.3 (https://assets.thermofisher.com/TFS-Assets/LSG/manuals/100079380_3500v3_3RUO_UG.pdf) for details regarding the Thermo Fisher Cloud app.
If you are using Data Collection Software v3.1, you would have to manually upload your data into the Thermo Fisher Cloud app from your data folder on the computer.
For Research Use Only. Not for use in diagnostic procedures.