missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ GeneScan™ 1000 ROX™ dye Size Standard

Catalog No. 401098
Encompass
Change view
Click to view available options
Quantity:
400 Reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
401098 400 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 401098 Supplier Applied Biosystems™ Supplier No. 401098

Includes

This kit contains the size standard

The GeneScan™ 1000 ROX™ Size Standard is an internal lane size standard developed for use with Applied Biosystems™ fluorescence-based DNA electrophoresis systems.

The GeneScan™ 1000 ROX™ Size Standard is an internal lane size standard developed for use with Applied Biosystems™ fluorescence-based DNA electrophoresis systems. The use of an internal lane size standard during electrophoresis enables automated data analysis and precise DNA fragment size comparisons between electrophoresis runs. GeneScan™ 1000 ROX™ Size Standard is designed for sizing DNA fragments in the 47–946 bp range and provides labeled fragments of 47, 51, 55, 82, 85 93 99, 126, 136, 262, 293, 317, 439, 557, 692, 695 and 946 bases. Each of the DNA fragments is labeled with ROX™ fluorophore, which results in a single peak when run under non-denaturing conditions.

Note: Under denaturing conditions using POP-4™ polymer, fragment lengths between 100 and 539 bp can be determined (the linear range of the separation with respect to length vs. scan number).

Order Info

Each

Specifications

Content And Storage This kit contains the size standard.
Store the kit at 2° to 8°C (Do not freeze).
For Use With (Equipment) 3730xl DNA Analyzer, 3730 DNA Analyzer, 3500 Series Genetic Analyzers, SeqStudio Flex Series Genetic Analyzers, SeqStudio Genetic Analyzer
For Use With (Application) Fragment analysis
Label or Dye ROX
No. of Reactions 400 Reactions
Product Line GeneScan, ROX
Product Type DNA Seqeuncing Size Standard
Quantity 400 Reactions
Shipping Condition Wet Ice
If my fragments are greater than 600 bp, can I use the GeneScan 1200 LIZ Dye, GeneScan 1000 ROX Dye, or GeneScan 2500 ROX Dye Size Standards?

For fragments that are greater than 600 bp, the GeneScan 1200 LIZ Dye Size Standard is recommended. The GeneScan 1000 ROX Dye, or GeneScan 2500 ROX Dye Size Standards are intended for non-denaturing applications only.

For Research Use Only. Not for use in diagnostic procedures.