missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BTX™ Genepaddles™ Electroporation Electrodes
Description
- Compatible with BTX Gemini X2 and ECM-2001 electroporation systems
- Two models to suit the size of the target area
- Gold plated, noninvasive
- Thickness: 1 mm
- Easily cleaned
- Life span is approximately 1500 pulses
- 450122 and 450123 require 45-0203 holder and 45-0216 cables (not included)
Specifications
Specifications
| For Use With (Application) | In-Vivo Gene Delivery, in-Vitro Embryo Gene Delivery |
| For Use With (Equipment) | ECM 830, ECM 2001+, Legacy ECM 2001, and Gemini X2 |
| Material | Gold-plated |
| Product Line | BTX Electroporators |
| Product Type | Genepaddles |
| Voltage | 200 V |
| Display | Color |
| Electrode Shape | Rectangular |
| Length (English) | 0.19 in. |
| Length (Metric) | 5 mm |
| Show More |
Intended for Research use only. Not for use in diagnostic or clinical procedures.