missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BTX™ Genepaddles™ Electroporation Electrodes
Genepaddles™ Electroporation Electrodes are paddle-style reusable electrodes suitable for both in vitro and in vivo gene delivery.
Supplier: BTX™ 45 0169
Description
- Compatible with BTX Gemini X2 and ECM-2001 electroporation systems
- Two models to suit the size of the target area
- Gold plated, noninvasive
- Thickness: 1 mm
- Easily cleaned
- Life span is approximately 1500 pulses
- 450122 and 450123 require 45-0203 holder and 45-0216 cables (not included)
Chemical Identifiers
| In-Vivo Gene Delivery, in-Vitro Embryo Gene Delivery | |
| Gold-plated | |
| 10 μsec. to 99 msec. in PBS | |
| 0.03 to 0.39 in. | |
| Non-sterile | |
| Square Wave |
| ECM 830, ECM 2001+, Legacy ECM 2001, and Gemini X2 | |
| 200 V | |
| 1 Ea. | |
| 1 to 10 mm | |
| Banana Plug to Micrograbber |
Specifications
| In-Vivo Gene Delivery, in-Vitro Embryo Gene Delivery | |
| Gold-plated | |
| Genepaddles Kit | |
| Rectangular | |
| 0.11 in. | |
| ∼200+ set of pulses | |
| 1 Ea. | |
| 1 to 10 mm | |
| 0.19 in. | |
| Non-sterile | |
| Square Wave |
| ECM 830, ECM 2001+, Legacy ECM 2001, and Gemini X2 | |
| BTX Electroporators | |
| 200 V | |
| Micrograbber cable, square post cable, banana plug and banana cable, genetrode holder, genepaddle | |
| 3 mm | |
| 10 μsec. to 99 msec. in PBS | |
| 0.03 to 0.39 in. | |
| Gold Plated, 3 x 5 mm | |
| 5 mm | |
| Banana Plug to Micrograbber |
Intended for Research use only. Not for use in diagnostic or clinical procedures.