missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ GeneMapper™ Software v4.0, client installation

Catalog No. 4440914
Encompass
Change view
Click to view available options
Quantity:
1 License
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
4440914 1 License
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 4440914 Supplier Applied Biosystems™ Supplier No. 4440914

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This client version of GeneMapper™ Software v4 installs the client configuration (user interface) only.

This client version of GeneMapper™ Software v4 installs the client configuration (user interface) only. Users must install GeneMapper™ Software 4, full installation, prior to installing the client version, which should be installed on a computer separate from that of the full version.

GeneMapper™ Software is a flexible fragment analysis software package that provides quality DNA sizing and allele calls for all Applied Biosystems™ genetic analyzers. This software is designed to support multi-application functionality, including analysis of amplified fragment length polymorphism (AFLP), loss of heterozygosity (LOH), microsatellites, and SNP genotyping.

Features include:

  • Multi-user, client-server deployment (through Oracle 11g RDBMS)
  • Increased data processing efficiency through remote auto-analysis and command line operation
  • Security and audit features to help you meet 21CFR11 requirements
  • Organize, manage, and batch-process your data with Report Manager and Database Dashboard
  • Automatically evaluate and reduce data review time with Process Quality Values (PQVs) for high-throughput genotyping

The software supports a wide variety of applications on the Applied Biosystems™ 3500, 3500xL, 3730, 3730xl, 3130, 3130xl, 3100, 3100–Avant, and 310 instrument systems.

Related Links:

Specifications

Content And Storage Software/CD
For Use With (Equipment) 310 Genetic Analyzer, 3100 Genetic Analyzer, 3100-Avant Genetic Analyzer, 3500xL Dx Genetic Analyzer, 3700 Genetic Analyzer, 3500 Dx Genetic Analyzer, 3730xl DNA Analyzer, 3130 Genetic Analyzer, 3730 DNA Analyzer, 3500xL Genetic Analyzer, 3500 Genetic Analyzer
For Use With (Application) Sequencing
Operating System Windows™ XP Pro SP2, Windows™ XP Pro SP1
Product Line GeneMapper
Product Type Software
Quantity 1 License
Software 1 Client License
Shipping Condition Room Temperature
How can I import Project Information into GeneMapper Manager?

Go to the Tools menu and select GeneMapper Manager. Click on the tab for the file(s) you want to import. Click on Import at the bottom of the window and select the files to be imported. Note: Multiple projects can be selected for importing at one time, but all other tabs are one at a time. Once imported, the new files will appear in the list. When finished, click Done in the lower right corner.

How can I back up my data files for DNA sequencing and fragment analysis applications, with the following extension, .ab1 or .fsa, on the capillary electrophoresis instruments?

The location of the sample files, .ab1 or .fsa, will either be in the Runs folder (D:/AppliedBio/310/Runs) on the ABI PRISM 310 Genetic Analyzer system or it will be determined by the Results Group in the Data Collection Software on the multi-capillary platforms. If you do not know where that location is, you can go to the Data Collection Software for your instrument and go to the Results Group section (top left corner in Unified Data Collection; for 3500 Data Collection, go to Library and in the Manage section, select Results Group), open the Results Group, and check the Sample File Folder location.
Save the files to the storage media of your choice (CD, DVD, network drive, flash drive).

Do you have a manual uninstall procedure for Full Install configurations on a stand-alone computer for GeneMapper Software v.4.0?

Please download this PDF (http://www.thermofisher.com/content/dam/LifeTech/Documents/PDFs/GeneMapper-Software-v.4-uninstall-full-install.pdf) and follow the steps listed to uninstall GeneMapper Software v.4.0.

I need to uninstall GeneMapper Software v.4.0 Client because of failed installation. How can I do this for Client Install configurations on a stand-alone computer?

Please download this PDF (http://www.thermofisher.com/content/dam/LifeTech/Documents/PDFs/GeneMapper-Software-v4-uninstall-client-install.pdf) and follow the steps listed to uninstall GeneMapper Software v.4.0 from your stand-alone computer.

How can I set up auto-analysis for data in GeneMapper with the 3500 Data Collection Software version 3.x?

Beginning with 3500 Data Collection Software version 3, the 3500 Data Collection Software does not support automatic secondary analysis. This is also indicated in the software release notes. To perform secondary data analysis, you will need to manually import the .fsa file into the GeneMapper software. For optimal performance, we recommend that you install the GeneMapper Software on a stand-alone computer that is not part of the 3500 Genetic Analyzer system.

Can the GM4 license (GeneMapper Software v4.0, client installation, Cat. No. 4440914) be used to register for GM5 (GeneMapper Software 5, client installation, Cat. No. A38892) or GM6 (GeneMapper Software 6, client installation, Cat. No. 4475074) copies?

No. You must get a GM5/GM6 license separately.

We are upgrading to the Windows 10 operating system. What are the compatible capillary electrophoresis secondary analysis software with Windows 10?

The following capillary electrophoresis secondary analysis software are compatible with Windows 10 (and also Windows 7):

-Sequencing Analysis Software v7
-SeqScape Software v4
-Variant Reporter Software v3
-GeneMapper Software v6
-Minor Variant Finder v1.2
-MicrobeBridge v1.1

We are upgrading to the Windows 10 operating system. Can I install GeneMapper Software v5 on that?

GeneMapper Software v5 is not Windows 10 compatible. GeneMapper Software v6 is compatible with Windows 10 and also Windows 7.

Where do I get the latest Applied Biosystems Capillary Electrophoresis Software patches?

Please visit our Applied Biosystems Genetic Analyzer Freeware and Patches website: https://www.thermofisher.com/sangerpatches

How can I back up my Data Files or Sample Files?

When a GeneMapper Project is created, the sample files are not imported into GeneMapper Software, the location of the sample files is mapped within the project and edits are done to the project, leaving the raw data intact. This means that backing up a project will not also back up the files associated with the project. They will have to be backed up separately and reassociated to the project again if they are moved or if the software is reinstalled.

The location of the sample files will either be in the Runs folder (D:/AppliedBio/310/Runs) on the ABI PRISM 310 Genetic Analyzer system or it will be determined by the Results Group in the Data Collection Software on the multi-capillary platforms. If you do not know where that location is, you can go to the Data Collection Software for your instrument and go to the Results Group section (top left corner in Unified Data Collection; for 3500 Data Collection, go to Library and in the Manage section, select Results Group), open the Results Group, and check the Sample File Folder location.

Save the files to the storage media of your choice (CD, DVD, network drive, flash drive). If you are moving projects to a new system or reinstalling GeneMapper Software because of a hard drive failure, then you can move the sample files to the new location or new drive. When you open the project in GeneMapper Software, go to File Associate Samples and browse to the folder containing the samples for the project.

How can I back up the data found in GeneMapper Manager?

To export project information, follow the steps outlined below:

1. Go to the Tools menu and select GeneMapper Manager.
2. Click on the tab that contains the file(s) you want to back up or export.
3. Select the file(s) that you want to export. Once they are selected, click on Export at the bottom of the window. (Projects can be selected by group, but the Save display will come up for each selected Project. In all of the other tabs, the files need to be exported one at a time).
4. Browse to the location where you want to save the files and click on Export to save it.
5. Save the files to the storage media of your choice (CD, DVD, network drive, flash drive). If the GeneMapper Software needs to be reinstalled or if you wish to put the information on another system, you can go to the GeneMapper Manager, select the tab into which you wish to import, click on Import and import the files. Once the files are saved, you can delete any old files or projects to conserve or free up space in the database.

What information should I back up when using GeneMapper Software?

It is useful to back up data in order to protect yourself from data loss, clearing out the Oracle database to keep it from filling up, and/or for sending data to Technical Support for troubleshooting.

In GeneMapper Software, there are 3 locations where you will need to back up data:

1. Panel Manager - contains information on your Panels, Markers, and Bins.
2. GeneMapper Manager - contains information pertaining to the Project itself - Projects, Analysis Methods, Size Standard Definitions, Table Settings, Plot Settings, Matrices (ABI PRISM 310 Genetic Analyzer Only), Cluster Plot Settings, SNP Sets, and Report Settings.
3. Data Files - sample data (.fsa files) that were used in the Project.

Note: Older versions of GeneMapper Software contained an Archive function that would back up the entire database at once. However, the archive could only be reimported into the same instance of GeneMapper Software. If you had a problem that required reinstallation of GeneMapper Software, it would be considered a new instance of the software and the Archive could not be imported into it.

I tried to launch GeneMapper Software and am getting a "Cannot connect to the Database" error message. The software is installed in a Client/Server configuration (Client and Server are on different computers). What should I do?

Please download this PDF (https://www.thermofisher.com/content/dam/LifeTech/Documents/PDFs/GeneMapper-Software-troubleshooting-client-install.pdf) for directions on how to resolve this error.

I tried to launch GeneMapper Software and am getting a "Cannot connect to the Database" error message. The software is installed on a stand-alone configuration, where the computer is not connected to the instrument. What should I do?

Please download this PDF (https://www.thermofisher.com/content/dam/LifeTech/Documents/PDFs/GeneMapper-Software-troubleshooting-full-install.pdf) for directions on how to resolve this error.

I tried to launch GeneMapper Software and am getting a "Cannot connect to the Database" error message. The software is installed on the instrument computer, where my computer is connected to the instrument (Client and Server on same computer). What should I do?

Please download this PDF (https://www.thermofisher.com/content/dam/LifeTech/Documents/PDFs/GeneMapper-Software-troubleshooting-full-install-instrument.pdf) for directions on how to resolve this error.

My Oracle database became corrupted and I have to reinstall the GeneMapper Software. However, when I try to uninstall using Add/Remove programs, it won't work. How do I manually uninstall GeneMapper Software?

Before uninstalling, make sure you have your GeneMapper CD/DVD and your registration code for reinstallation. Follow the uninstall instructions for your version of GeneMapper Software. Before attempting to reinstall, verify that you are logged in locally as the Administrator, that your antivirus Software is either off or that you have suspended activity during installation, and that your TCP/IP is installed.

When installing GeneMapper Software, the installation stalled at 99%. What do I do next?

Follow the uninstall instructions for your version of GeneMapper Software. Before attempting to reinstall, verify that you are logged in locally as the Administrator, that your antivirus software is either off or that you have suspended activity during installation, and that your TCP/IP is installed.

We are upgrading to the Windows 7 operating system. What version of GeneMapper Software is compatible with that?

GeneMapper Software 5 has been extensively tested on Windows 7 SP1 (32-bit version). While there are some indications that software will run normally on the 64-bit version of Windows 7 SP1, the 64-bit configuration is not officially supported. It is strongly recommended that this software be run on Windows 7 SP1 (32-bit). For a datasheet on GeneMapper Software 5, please click here (https://www.thermofisher.com/content/dam/LifeTech/Documents/PDFs/GeneMapper-Software-5-datasheet.pdf).

If installing on the instrument computer, the update will require a new computer and upgrading of the Data Collection Software as well.

I have a copy of GeneMapper Software v.4.0 and want to know what the computer requirements are.

The computer requirements are as follows:

-Minimum: 2.4 GHz CPU or faster
-1 GB RAM
-120 GB hard disk space*

*Hard drive requirements will vary depending on how much space is allocated to the Oracle Database. Having a large hard drive will allow for database expansion should the database fill up.

For the operating system, GeneMapper Software v.4.0 requires Windows XP Professional, service pack 1 or 2, or Windows 2000, service pack 4. Please click here (https://www.thermofisher.com/content/dam/LifeTech/Documents/PDFs/GeneMapper-Software-v4-datasheet.pdf) for the GeneMapper Software v.4.0 datasheet

We are upgrading to the Windows 7 operating system. Can I install GeneMapper Software v.4.0 on that?

Unfortunately, GeneMapper Software v.4.0 is not Windows 7 compatible. Please click here (http://www.thermofisher.com/content/dam/LifeTech/Documents/PDFs/GeneMapper-Software-v4-datasheet.pdf) for the GeneMapper Software v.4.0 datasheet.


For Research Use Only. Not for use in diagnostic procedures.