missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ GeneJET Stabilized and Fresh Whole Blood RNA Kit

Catalog No. FERK0871
Change view
Click to view available options
Quantity:
50 preps
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FERK0871 50 preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FERK0871 Supplier Thermo Scientific™ Supplier No. K0871

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Thermo Scientific GeneJET™ Stabilized and Fresh Whole Blood RNA Kit is designed for rapid and efficient purification of high quality total RNA from various whole blood samples: fresh, frozen, or human blood samples stabilized with commercial stabilizers such as Tempus™ and PAXgene™.

The GeneJET™ Stabilized and Fresh Whole Blood RNA Kit is designed for rapid and efficient purification of high quality total RNA from various whole blood samples: fresh, frozen, or human blood samples stabilized with commercial stabilizers such as Tempus™ and PAXgene™.

The kit utilizes silica-based membrane technology in the form of a convenient spin column, eliminating the need for expensive resins, toxic phenol-chloroform extractions, or time-consuming alcohol precipitation. The kit includes a DNase l for the enzymatic removal of genomic DNA. RNA can be isolated from whole blood in less than 30 minutes (including cell lysis and DNase l treatment).

Thermo Scientific GeneJET™ Stabilized and Fresh Whole Blood RNA Kit is designed for rapid and efficient purification of high quality total RNA from various whole blood samples: fresh, frozen, or human blood samples stabilized with commercial stabilizers such as Tempus™ and PAXgene™.

The kit utilizes silica-based membrane technology in the form of a convenient spin column, eliminating the need for expensive resins, toxic phenol-chloroform extractions, or time-consuming alcohol precipitation. The kit includes a DNase l for the enzymatic removal of genomic DNA. RNA can be isolated from whole blood in less than 30 minutes (including cell lysis and DNase l treatment).

Highlights

  • Versatile—compatible with fresh and frozen blood, buffy coat, as well as blood collected in PAXgene™and Tempus™ Tubes
  • Convenient—specially optimized Lysis Buffer is suitable for RNA stabilization up to 48 h
  • Efficient—up to 4 μg of total RNA from 500 μL of healthy human blood
  • Pure—isolates intact total RNA free of any RT-qPCR inhibitors

Applications

Isolated total RNA can be used in:

  • RT-PCR
  • RT-qPCR
  • Northern Blotting

Includes

  • Lysis Buffer (R2841)
  • Wash Buffer 1 (conc.)(R2851)
  • Wash Buffer 2 (conc.)(R2861)
  • DNase l (lyophilized)
  • DNase I Reconstitution Buffer
  • 2x DNase I Buffer
  • Manganese Chloride Solution
  • Water, nuclease-free
  • GeneJET RNA Purification Columns pre-assembled with Collection Tubes
  • Collection Tubes, 2 mL
  • Collection Tubes, 1.5 mL

Specifications

Concentration 2X
Content And Storage For better long-term performance, it is recommended to store the purification columns at 2°C-8°C.
Format Spin Column
Isolation Technology Spin Column
Purification Time 30 min.
Sample Type Blood
High-throughput Compatibility Not High-throughput Compatible (Manual)
For Use With (Application) Real-Time Quantitative PCR (qPCR), Reverse Transcriptase PCR (RT-PCR), Nuclease Protection Assays, Nucleic Acid Labeling, Northern Blotting, cDNA Library Construction
No. of Reactions 50 Preps
Product Line GeneJET
Quantity 50 preps
Target Total RNA
Show More Show Less
Can I use water instead of Elution Buffer in the elution step of GeneJET and MagJET nucleic acid purification kits?

We would highly recommend using the provided Elution Buffer in the elution step for best results. You may use nuclease-free water; however, it may affect the yield of the nucleic acids during purification and their stability during storage.

Is any GeneJET kit suitable for RNA purification from formalin-fixed paraffin-embedded FFPE tissues?

We cannot recommend GeneJET kits for RNA purification from FFPE tissues. In case of genomic DNA purififcation from formalin-fixed paraffin-embedded samples, we recommend GeneJET FFPE DNA Purification Kit.

Are MagJET and GeneJET blood nucleic acid purification kits compatible with animal blood?

Yes, the kits are compatible with animal blood for DNA/RNA purification.

Can I use a vacuum manifold with GeneJET kits?

The GeneJET spin columns are compatible with vacuum manifolds of standard size (e.g. Qiagen, Sigma-Aldrich, Promega). However, not all GeneJET kits are supported with a vacuum protocol. Please refer to the particular kit protocol for the detailed procedure.

At what temperature should I perform centrifugation steps in the GeneJET kits?

We recommend performing centrifugation steps at room temperature when the temperature is not specified.

Are GeneJET spin columns or MagJET magnetic beads available separately?

No, we do not offer GeneJET columns or MagJET magnetic beads separately at this time.

Does the GeneJET Stabilized and Fresh Whole Blood RNA Kit isolate miRNA?

This kit is not suitable for purification of small RNAs.

Do the GeneJET RNA purification kits remove genomic DNA?

These kits will remove the majority of genomic DNA, however trace amounts may still remain. Therefore, for applications that are very sensitive to genomic DNA, we recommend you treat the purified RNA sample with DNase I (Cat. No. EN0521), RapidOut DNA Removal Kit (Cat. No. K2981), or dsDNase (Cat. No. EN0771).


For Research Use Only. Not for use in diagnostic procedures.