missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ GeneClean Kit

Catalog No. 111001200
Change view
Click to view available options
No. of Reactions:
200 Preps
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Reactions
111001200 200 Preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111001200 Supplier MP Biomedicals™ Supplier No. 111001200

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

GeneClean™ Kit, Solution or Gels

  • The GENECLEAN™ Kit includes Glassmilk™ slurry that allows flexibility of reaction size and volume.
  • The GENECLEAN™ kit simplifies the process of purifying DNA into three easy steps: Bind,Wash and Elute.
  • Ethanol precipitation is never required and purified DNA is immediately ready for a wide variety of downstream applications.
  • Isolate DNA from TAE buffered agarose gels and solutions
  • The GENECLEAN™ Kit is used to purify fragments of DNA from 200 bp to 20 kb from solutions and agarose gels.
  • Examples of applications are desalting, elimination of proteins from enzymatic reactions, removal of primers and unincorporated nucleotides from enzymatic reactions, separation of linearized from uncut vector and isolation of PCR products away from genomic DNA.

Specifications

Concentration New Wash/Ethanol Concentration must be ∽ 50%
Content And Storage Store at Room Temperature (15-30°C). NaI solution must be kept away from light and open air sources to prevent oxidation. NaI is packaged under nitrogengas to further reduce chance of oxidation. Shelf life may be extended by storing at 4°C.
For Use With (Application) Examples of applications are desalting, elimination of proteins from enzymatic reactions, removal of primers and unincorporated nucleotides from enzymatic reactions, separation of linearized from uncut vector and isolation of PCR products away fromgenomic DNA.
No. of Reactions 200 Preps
pH Above 500 bp: 4.5 M NaI pH 7.0 - 7.4; Between 200 - 500 bp: 4.5 M NaI pH 6.0; NEW Wash: pH Range: 7.4 - 8.0
Product Type Kits and Assays
Form Solution or Gels