missing translation for 'onlineSavingsMsg'
Learn More

GemPure™ Select Water for Injection (WFI) Quality Water (USP Grade), GeminiBio™

Catalog No. 11852127
Change view
Click to view available options
Quantity:
6 X 500 mL
10 L
20 L
6 x 1 L
100 L
200 L
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11852127 10 L
11852126 6 x 1 L
11852118 20 L
11852125 100 L
11852115 200 L
11852119 6 X 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
For further manufacturing or research use only. Not for direct Therapeutic use.
Catalog No. 11852127 Supplier Gemini Bio Products Supplier No. 901001011
Only null left
For further manufacturing or research use only. Not for direct Therapeutic use.

GemPure™ Select Water for Injection (WFI) Quality Water (USP Grade) meets USP monograph requirements to support upstream and downstream bioprocessing from development through commercial production.

Designed for consistency and reliability, this grade supports critical bioprocess workflows where USP compliance and controlled production are required.

  • Meets USP monograph requirements for Water for Injection packaged in bulk
  • Supports upstream and downstream bioprocessing from development to commercial production
  • Manufactured in an ISO 13485:2016 and 21 CFR 820 compliant facility
  • Designed for consistent purity and reliable performance
  • Available in 500 mL, 1 L, 10 L, 20 L, 100 L, 200 L, and custom sizes

Suited for

  • Manufacturing workflows requiring USP-compliant WFI
  • Programs progressing from development into commercial production
  • Customers seeking scalable, bulk-packaged WFI options

Chemical Identifiers

CAS 7732-18-5

Specifications

pH 5.0 to 7.0
Color Clear and Colorless
Conductivity <5.0 μS/cm
DNase DNase-free
Endotoxin <0.25 EU/mL
EU Ph Reference 169
Molecular Weight (g/mol) 18
ChEBI 15377
Percent Purity 0.99999
Grade USP
Concentration or Composition (by Analyte or Components) H2O
Sterility Sterile
Filtered Through 0.2 μM Sterilization Grade Filter
Density 1 g/mL
Certificate of Analysis Yes
Quantity 10 L
Purification Method Filtration RO/DI
Shipping Condition Ambient protect from freezing
Chemical Name or Material Water, Water For Injection Quality
Show More Show Less
ShelfLife 24 Months
EINECSNumber 231-791-2
Recommended Storage 2°C to 30°C