missing translation for 'onlineSavingsMsg'
Learn More

Gemfibrozil 98.0+%, TCI America™

Catalog No. G03685G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G03685G 5 g
G036825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. G03685G Supplier TCI America Supplier No. G03685G
Only null left

Chemical Identifiers

CAS 25812-30-0
Molecular Formula C15H22O3
Molecular Weight (g/mol) 250.34
MDL Number MFCD00079335
InChI Key HEMJJKBWTPKOJG-UHFFFAOYSA-N
Synonym gemfibrozil, lopid, 5-2,5-dimethylphenoxy-2,2-dimethylpentanoic acid, jezil, decrelip, lipur, bolutol, apo-gemfibrozil, gemfibromax, cholespid
PubChem CID 3463
ChEBI CHEBI:5296
IUPAC Name 5-(2,5-dimethylphenoxy)-2,2-dimethylpentanoic acid
SMILES CC1=CC=C(C)C(OCCCC(C)(C)C(O)=O)=C1

Specifications

Melting Point 61°C
Color White
Boiling Point 159°C
Quantity 5 g
Physical Form Crystal-Powder at 20°C
Formula Weight 250.34
Percent Purity ≥98.0% (GC,T)
Chemical Name or Material Gemfibrozil
RTECSNumber YV7120000
TSCA Yes