Learn More
Gemcitabine hydrochloride, 98%, Thermo Scientific™
Chemical Identifiers
| CAS | 122111-03-9 |
|---|---|
| Molecular Formula | C9H12ClF2N3O4 |
| Molecular Weight (g/mol) | 299.66 |
| InChI Key | OKKDEIYWILRZIA-OSZBKLCCSA-N |
| IUPAC Name | hydrogen 4-amino-1-[(2R,4R,5R)-3,3-difluoro-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-1,2-dihydropyrimidin-2-one chloride |
| SMILES | [H+].[Cl-].NC1=NC(=O)N(C=C1)[C@@H]1O[C@H](CO)[C@@H](O)C1(F)F |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AC461060010
|
Thermo Scientific Chemicals
461060010 |
1 g |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
AC461060050
|
Thermo Scientific Chemicals
461060050 |
5 g |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 122111-03-9 | |
| 299.66 | |
| hydrogen 4-amino-1-[(2R,4R,5R)-3,3-difluoro-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-1,2-dihydropyrimidin-2-one chloride |
| C9H12ClF2N3O4 | |
| OKKDEIYWILRZIA-OSZBKLCCSA-N | |
| [H+].[Cl-].NC1=NC(=O)N(C=C1)[C@@H]1O[C@H](CO)[C@@H](O)C1(F)F |
Specifications
| 122111-03-9 | |
| C9H12ClF2N3O4 | |
| Solubility aq. soln.: soluble. Other solubilities: slightly soluble in methanol, practically insoluble in ethanol and other polar, organic solvents | |
| [H+].[Cl-].NC1=NC(=O)N(C=C1)[C@@H]1O[C@H](CO)[C@@H](O)C1(F)F | |
| 299.66 | |
| Gemcitabine hydrochloride |
| 97.5 % min. (HPLC) | |
| 1 g | |
| OKKDEIYWILRZIA-OSZBKLCCSA-N | |
| hydrogen 4-amino-1-[(2R,4R,5R)-3,3-difluoro-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-1,2-dihydropyrimidin-2-one chloride | |
| 299.66 |
Safety and Handling
GHS H Statement Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause genetic defects.
May damage fertility or the unborn child.
GHS P Statement IF ON SKIN: Wash with plenty of soap and water.
Call a POISON CENTER or doctor/physician if you feel unwell.
If eye irritation persists: Get medical advice/attention.
Obtain special instructions before use.
Wear protective gloves/protective clothing/eye protection/face protection.
GHS Signal Word: Danger
RUO â Research Use Only