missing translation for 'onlineSavingsMsg'
Learn More

Gemcitabine hydrochloride, 98%

Catalog No. AC461060010
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC461060010 1 g
AC461060050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC461060010 Supplier Thermo Scientific Chemicals Supplier No. 461060010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 122111-03-9 | C9H12ClF2N3O4 | 299.66 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 122111-03-9
Molecular Formula C9H12ClF2N3O4
Molecular Weight (g/mol) 299.66
InChI Key OKKDEIYWILRZIA-OSZBKLCCSA-N
IUPAC Name hydrogen 4-amino-1-[(2R,4R,5R)-3,3-difluoro-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-1,2-dihydropyrimidin-2-one chloride
SMILES [H+].[Cl-].NC1=NC(=O)N(C=C1)[C@@H]1O[C@H](CO)[C@@H](O)C1(F)F

Specifications

Assay Percent Range 97.5 % min. (HPLC)
Quantity 1 g
Solubility Information Solubility aq. soln.: soluble. Other solubilities: slightly soluble in methanol, practically insoluble in ethanol and other polar, organic solvents
Formula Weight 299.66
Chemical Name or Material Gemcitabine hydrochloride
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement Harmful in contact with skin.

Causes skin irritation.

Causes serious eye irritation.

May cause genetic defects.

May damage fertility or the unborn child.

Precautionary Statement GHS P Statement IF ON SKIN: Wash with plenty of soap and water.

Call a POISON CENTER or doctor/physician if you feel unwell.

If eye irritation persists: Get medical advice/attention.

Obtain special instructions before use.

Wear protective gloves/protective clothing/eye protection/face protection.

RUO – Research Use Only