missing translation for 'onlineSavingsMsg'
Learn More

Gemcitabine, 98%

Catalog No. AC456890010
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC456890010 1 g
AC456890050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC456890010 Supplier Thermo Scientific Chemicals Supplier No. 456890010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 95058-81-4 | C9H11F2N3O4 | 263.2 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 95058-81-4
Molecular Formula C9H11F2N3O4
Molecular Weight (g/mol) 263.2
InChI Key SDUQYLNIPVEERB-QPPQHZFASA-N
Synonym gemcitabine, 2',2'-difluorodeoxycytidine, gemcitabinum, gamcitabine, gemcitabina, gemcitabine hcl, dfdc, 2'-deoxy-2',2'-difluorocytidine, folfugem, gemcel
PubChem CID 60750
ChEBI CHEBI:175901
IUPAC Name 4-amino-1-[(2R,4R,5R)-3,3-difluoro-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]pyrimidin-2-one
SMILES C1=CN(C(=O)N=C1N)C2C(C(C(O2)CO)O)(F)F

Specifications

Melting Point 292.0°C
Loss on Drying 2% max.
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 263.2
Percent Purity 98%
Chemical Name or Material Gemcitabine
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
Suspected of damaging fertility.
Suspected of damaging the unborn child.
May cause damage to organs through prolonged or repeated exposure if inhaled.<br
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Take off contaminated clothing and wash before reuse.
IF IN EYES: Rinse cautiously with water

RUO – Research Use Only