CAS RN 95058-81-4
IUPAC Name:
4-amino-1-[(2R,4R,5R)-3,3-difluoro-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-1,2-dihydropyrimidin-2-one
Synonyms:
gemcitabine gamcitabine gemtro GEO gemzar gemcin DFdCyd gemcitabinum gemcitabina gemcitabina [inn-spanish]
Molecular Weight (g/mol):
263.2
Molecular Formula:
C9H11F2N3O4
InChi Key:
SDUQYLNIPVEERB-QPPQHZFASA-N
SMILES:
NC1=NC(=O)N(C=C1)[C@@H]1O[C@H](CO)[C@@H](O)C1(F)F
1
–
1
of
1
results
1
Promotions Available
Gemcitabine, 98%
Gemcitabine, 98%
CAS: 95058-81-4 Molecular Formula: C9H11F2N3O4 Molecular Weight (g/mol): 263.2 InChI Key: SDUQYLNIPVEERB-QPPQHZFASA-N Synonym: gemcitabine,2',2'-difluorodeoxycytidine,gemcitabinum,gamcitabine,gemcitabina,gemcitabine hcl,dfdc,2'-deoxy-2',2'-difluorocytidine,folfugem,gemcel PubChem CID: 60750 ChEBI: CHEBI:175901 IUPAC Name: 4-amino-1-[(2R,4R,5R)-3,3-difluoro-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]pyrimidin-2-one SMILES: C1=CN(C(=O)N=C1N)C2C(C(C(O2)CO)O)(F)F