missing translation for 'onlineSavingsMsg'
Learn More

GeminiBio™ GemCell™ Plus Xeno-Free, World Grade

Catalog No. 11852109
Change view
Click to view available options
Sterilization Method:
Gamma-irradiated
Sequential-filtered, 100 nm
Format:
Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sterilization Method Format
11852109 Sequential-filtered, 100 nm Bottle
11852104 Gamma-irradiated Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
For further manufacturing or research use only. Not for direct Therapeutic use.
Catalog No. 11852109 Supplier GeminiBio™ Supplier No. 100916100
For further manufacturing or research use only. Not for direct Therapeutic use.

Collected from a controlled pool of up to 16 healthy male AB donors and defibrinated using therapeutic-grade recombinant human thrombin. Xeno-Free and extensively tested to meet EU requirements for clinical and commercial use.

GemCell™ Plus Xeno-Free, World Grade is designed for global programs requiring tighter donor control and enhanced traceability. A capped donor pool and Xeno-Free processing support alignment with EU-focused regulatory strategies and region-specific qualification requirements.

  • Collected from a controlled pool of up to 16 healthy male AB donors
  • Defibrinated using therapeutic-grade recombinant human thrombin
  • Manufactured through a Xeno-Free process
  • Extensively tested to support clinical and commercial use
  • Controlled donor strategy enhances traceability and lot consistency

Suited for:

  • Global or EU-aligned programs requiring enhanced donor control
  • Applications prioritizing traceability and consistency
  • Customers balancing regulatory alignment with defined supply strategies

Specifications

Form Liquid
Format Bottle
Product Type Human Serum
Quantity 100 mL
Sterility Sterile-filtered
Sterilization Method Sequential-filtered, 100 nm