missing translation for 'onlineSavingsMsg'
Learn More

Gellan Gum Powder, MP Biomedicals™

Catalog No. p-7083173
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
100 g
Catalog No. Quantity
MP021801061 100 g
1 options

Catalog No. MP021801061

Supplier: MP Biomedicals Inc 2180106.1

Only null left

Used as a gelling agent in place of agar in microbiological media. Gellan Gum Powder, MP Biomedicals is a water-soluble polysaccharide produced by Pseudomonas elodea, a bacterium. It is used in a variety of immobilization matricies and as a food additive,

Chemical Identifiers

CAS 71010-52-1
Molecular Formula (C24H38O20)A
Synonym Agar substitute gelling agent

Specifications

Quantity 100 g
Physical Form Powder
Recommended Storage Store at Room Temperature (15–30°C).