accessibility menu, dialog, popup

UserName

CAS RN 71010-52-1

IUPAC Name: —
Synonyms: —
Molecular Weight (g/mol): —
Molecular Formula: (C24H38O20)A
InChi Key: —
SMILES: [H][C@@]1(-*)O[C@@H](C)[C@H](O[C@]2([H])O[C@H](CO)[C@@H](O[C@]3([H])O[C@@H]([C@@H](O[C@]4([H])O[C@H](CO)[C@@H](O)[C@H](O-*)[C@H]4O)[C@H](O)[C@H]3O)C(O)=O)[C@H](O)[C@H]2O)[C@@H](O)[C@H]1O
Quantity 
  • (1)
  • (1)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)