missing translation for 'onlineSavingsMsg'
Learn More

Gelatin, 250 Bloom, Powder, NF, Spectrum™ Chemical

GE105, 9000-70-8

$462.69 - $17549.34

Chemical Identifiers

CAS 9000-70-8
MDL Number MFCD00081638
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
18602891
View Documents
Spectrum Chemical Mfg Cor
GE105500GM
500 g Poly Bottle
Each for $462.69
Only null left
 
18602892
View Documents
Spectrum Chemical Mfg Cor
GE10525KG
2.5 kg Poly Pail
Each for $1,495.66
Only null left
 
18602893
View Documents
Spectrum Chemical Mfg Cor
GE10512KG
12 kg Fiber Drum
Each for $4,234.60
Only null left
 
18602894
View Documents
Spectrum Chemical Mfg Cor
GE10545KGBL
45 kg Fiber Drum N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Spectrum™ Chemical Gelatin, 250 Bloom, Powder, NF is used pharmaceutical formulations as thickening agent and emulsifier. All Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications

Chemical Identifiers

9000-70-8
MFCD00081638

Specifications

9000-70-8
15%
MFCD00081638
NF
100%
Poly Bottle
500 g
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'