missing translation for 'onlineSavingsMsg'
Learn More

Gelatin, 250 Bloom, Powder, NF, Spectrum™ Chemical

GE105, 9000-70-8

Supplier:  Spectrum Chemical Mfg Cor GE10525KG

 View more versions of this product

Catalog No. 18602892


Only null left
Explore available promotions

Description

Description

Spectrum™ Chemical Gelatin, 250 Bloom, Powder, NF is used pharmaceutical formulations as thickening agent and emulsifier. All Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications

Specifications

9000-70-8
15%
MFCD00081638
NF
100%
Poly Pail
2.5 kg

Chemical Identifiers

9000-70-8
MFCD00081638
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions