missing translation for 'onlineSavingsMsg'
Learn More

FUJIFILM Wako Gel Clot Reaction Tubes

Endotoxin-free gel clot reaction tubes made of borosilicate glass

$288.50 - $1,267.29

Specifications

Format Borosilicate Glass Test Tubes
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
12333029
View Documents Promotion Details
FUJIFILM Wako
CT1075
200 Tubes
Each for $288.50
Only null left
 
12333030
View Documents Promotion Details
FUJIFILM Wako
CT1075B
1250 Tubes
Each for $1,267.29
Only null left
 
Description

Description

  • Endotoxin-free test tubes (<0.001 EU/tube)
  • Endotoxin-free tubes depyrogenated at 250 °C
  • Made with borosilicate glass
  • Designed for gel clot testing
  • Dimensions: 10 x 75 mm
Specifications

Specifications

Borosilicate Glass Test Tubes
Videos
SDS
Documents

Documents