missing translation for 'onlineSavingsMsg'
Learn More

FUJIFILM Wako Gel Clot Reaction Tubes

Catalog No. 12333029
Change view
Click to view available options
Quantity:
1250 Tubes
200 Tubes
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
12333029 200 Tubes
12333030 1250 Tubes
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 12333029 Supplier FUJIFILM Wako Supplier No. CT1075

Endotoxin-free gel clot reaction tubes made of borosilicate glass

  • Endotoxin-free test tubes (<0.001 EU/tube)
  • Endotoxin-free tubes depyrogenated at 250 °C
  • Made with borosilicate glass
  • Designed for gel clot testing
  • Dimensions: 10 x 75 mm

Specifications

Format Borosilicate Glass Test Tubes
Quantity 200 Tubes