missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GEF5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554293
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554293 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554293 Supplier Invitrogen™ Supplier No. PA554293
Only null left

Rabbit Polyclonal Antibody

Immunogen sequence: PAREPLRRTT PQQGASGPGR SPVGQARQPE KPSHLHLEKA SSWPHRRDSG RPPGDSSGQA VAPSEGANKH KGWS Highest antigen sequence identity to the following orthologs: Mouse - 46%, Rat - 45%.

This gene encodes a protein that interacts with thyroid hormone receptors, also known as hormone-dependent transcription factors. The gene product interacts specifically with the ligand binding domain. This gene is one of several that may play a role in early-stage non-small cell lung cancer.

Specifications

Antigen GEF5
Applications Immunocytochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ARHGEF5
Gene Accession No. Q12774
Gene Alias 2210412D05Rik; AA717842; ARH5; ARHGEF5; AW495314; DKFZp686N1969; Ephexin 3; Ephexin-3; GEF5; Guanine nucleotide regulatory protein TIM; oncogene TIM; P60; p60 TIM; Rho guanine nucleotide exchange factor (GEF) 5; Rho guanine nucleotide exchange factor 5; TIM; TIM1; Transforming immortalized mammary oncogene
Gene Symbols ARHGEF5
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human GEF5. Recombinant protein control fragment (Product #RP-92619).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7984
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less