missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GDNF Polyclonal Antibody

Sheep Polyclonal Antibody
Supplier: Invitrogen™ PA118367
Description
Reconstitute in 100 μL of sterile water. Centrifuge to remove any insoluble material. After reconstitution keep aliquots at -20 °C for a higher stability, and at 4 °C with an appropriate antibacterial agent. Glycerol (1:1) may be added for an additional stability. Avoid repetitive freeze/thaw cycles.
GDNF is a glycosylated, disulfide-bonded homodimer molecule. It was first discovered as a potent survival factor for midbrain dopaminergic neurons and was then shown to rescue these neurons in animal models of Parkinson's disease. GDNF is about 100 times more efficient survival factor for spinal motor neurons than the neurotrophins. GDNF is a neurotrophic factor that enhances survival and morphological differentiation of dopaminergic neurons and increases their high-affinity dopamine uptake.Chemical Identifiers
| GDNF | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Polyclonal | |
| Lyophilized | |
| IgG | |
| Human, Mouse, Rat | |
| GDNF | |
| AI385739; Astrocyte-derived trophic factor; ATF; ATF1; ATF2; Gdnf; glial cell derived neurotrophic factor; glial cell line derived neurotrophic factor; Glial cell line derived neutrophic factor; glial cell line-derived neurotrophic factor; gndf; HFB1-GDNF; hGDNF; H-GDNF; HSCR3; mGDNF; neurotrophic factor | |
| GDNF | |
| Primary |
| Conc. Not Determined | |
| Western Blot | |
| Unconjugated | |
| Sheep | |
| RUO | |
| Whole serum with no preservative | |
| P39905, P48540, Q07731 | |
| 14573, 25453, 2668 | |
| Recombinant human GDNF (aa 78-211). | |
| Antibody |
Specifications
| GDNF | |
| Polyclonal | |
| Unconjugated | |
| GDNF | |
| AI385739; Astrocyte-derived trophic factor; ATF; ATF1; ATF2; Gdnf; glial cell derived neurotrophic factor; glial cell line derived neurotrophic factor; Glial cell line derived neutrophic factor; glial cell line-derived neurotrophic factor; gndf; HFB1-GDNF; hGDNF; H-GDNF; HSCR3; mGDNF; neurotrophic factor | |
| Sheep | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Western Blot | |
| Conc. Not Determined | |
| Whole serum with no preservative | |
| P39905, P48540, Q07731 | |
| GDNF | |
| Recombinant human GDNF (aa 78-211). | |
| RUO | |
| 14573, 25453, 2668 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |