missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GDF3 Recombinant Rabbit Monoclonal Antibody (23GB5540)
Rabbit Recombinant Monoclonal Antibody
$590.92 - $1416.21
Specifications
| Antigen | GDF3 |
|---|---|
| Clone | 23GB5540 |
| Content And Storage | -20°C |
| Applications | Western Blot |
| Classification | Recombinant Monoclonal |
Description
GDF3 Recombinant Monoclonal Antibody for Western Blot
GDF3 is a member of the bone morphogenetic protein (BMP) family and the TGF-beta superfamily. This group of proteins is characterized by a polybasic proteolytic processing site which is cleaved to produce a mature protein containing seven conserved cysteine residues. The members of this family are regulators of cell growth and differentiation in both embryonic and adult tissues. The function of this protein is unknown, but expression studies suggest it may be involved in regulation of the adult lymphatic and erythroid systems and embryonic development.Specifications
| GDF3 | |
| -20°C | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Human, Mouse, Rat | |
| GDF3 | |
| C78318; ecat9; Gdf3; Gdf-3; growth differentiation factor 3; growth/differentiation factor 3; KFS3; MCOP7; MCOPCB6; RGD1564178; UNQ222/PRO248; VG-1-related protein 2; Vgr2; Vgr-2 | |
| GDF3 | |
| Primary | |
| Affinity chromatography |
| 23GB5540 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q07104, Q9NR23 | |
| 14562, 500311, 9573 | |
| A synthesized peptide derived from human GDF3 (300-364AA). | |
| Antibody |