missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GDF3 Polyclonal Antibody, PeproTech®, Invitrogen™

Catalog No. 500P23550UG
Encompass
Change view
Click to view available options
Quantity:
100 μg
50 μg
1 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
500P23550UG 50 μg
500P235100 100 μg
500P2351MG 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 500P23550UG Supplier Invitrogen™ Supplier No. 500P23550UG
Season of Savings
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Rabbit Polyclonal Antibody

AA Sequence of recombinant protein: AAIPVPKLSC KNLCHRHQLF INFRDLGWHK WIIAPKGFMA NYCHGECPFS LTISLNSSNY AFMQALMHAV DPEIPQAVCI PTKLSPISML YQDNNDNVIL RHYEDMVVDE CGCG. Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Human GDF-3. Anti-Human GDF-3-specific antibody was purified by affinity chromatography employing an immobilized Human GDF-3 matrix. Sandwich ELISA: To detect Human GDF-3 by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.5-2.0 μg/mL of this antibody is required. This antigen affinity purified antibody, in conjunction with PeproTech Biotinylated Anti-Human GDF-3 (500-P235Bt) as a detection antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant Human GDF-3. Western Blot: To detect Human GDF-3 by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant Human GDF-3 is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions.

GDF3 is a member of the bone morphogenetic protein (BMP) family and the TGF-beta superfamily. This group of proteins is characterized by a polybasic proteolytic processing site which is cleaved to produce a mature protein containing seven conserved cysteine residues. The members of this family are regulators of cell growth and differentiation in both embryonic and adult tissues. The function of this protein is unknown, but expression studies suggest it may be involved in regulation of the adult lymphatic and erythroid systems and embryonic development.

Specifications

Antigen GDF3
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.1-1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene GDF3
Gene Accession No. Q9NR23
Gene Alias C78318; ecat9; Gdf3; Gdf-3; growth differentiation factor 3; growth/differentiation factor 3; KFS3; MCOP7; MCOPCB6; RGD1564178; UNQ222/PRO248; VG-1-related protein 2; Vgr2; Vgr-2
Gene Symbols GDF3
Host Species Rabbit
Immunogen E.coli-derived Recombinant Human GDF-3.
Purification Method Antigen affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9573
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Show More Show Less