missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GCN2 Recombinant Rabbit Monoclonal Antibody (23GB5295)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA553881
Description
GCN2 Recombinant Monoclonal Antibody for Flow, ICC/IF, Western Blot
EIF2AK4 (GCN2) is a metabolic-stress sensing protein kinase that phosphorylates the alpha subunit of eukaryotic translation initiation factor-2 in response to low amino acid availability. It plays a role as an activator of the integrated stress response required for adapatation to amino acid starvation. Mutations in the gene lead to pulmonary vennocclusive disease 2.Chemical Identifiers
| GCN2 | |
| 0.7 mg/mL | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| D4A7V9, Q9P2K8 | |
| 114859, 440275 | |
| A synthesized peptide derived from human GCN2 (1-59AA). | |
| Antibody |
| 23GB5295 | |
| -20°C | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Human, Rat | |
| EIF2AK4 | |
| 2610011M03; EIF2AK4; eIF-2-alpha kinase GCN2; eukaryotic translation initiation factor 2 alpha kinase 4; eukaryotic translation initiation factor 2-alpha kinase 4; Gcn2; GCN2 eIF2alpha kinase; GCN2-like protein; general control nonderepressible 2; Kiaa1338; mGCN2; PVOD2 | |
| EIF2AK4 | |
| Primary | |
| Affinity chromatography |
Specifications
| GCN2 | |
| Recombinant Monoclonal | |
| 0.7 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| D4A7V9, Q9P2K8 | |
| EIF2AK4 | |
| A synthesized peptide derived from human GCN2 (1-59AA). | |
| 20 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB5295 | |
| Unconjugated | |
| EIF2AK4 | |
| 2610011M03; EIF2AK4; eIF-2-alpha kinase GCN2; eukaryotic translation initiation factor 2 alpha kinase 4; eukaryotic translation initiation factor 2-alpha kinase 4; Gcn2; GCN2 eIF2alpha kinase; GCN2-like protein; general control nonderepressible 2; Kiaa1338; mGCN2; PVOD2 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 114859, 440275 | |
| -20°C | |
| Liquid |