missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GCM1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA577188
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA577188 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA577188 Supplier Invitrogen™ Supplier No. PA577188

Rabbit Polyclonal Antibody

The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).

This gene encodes a DNA-binding protein with a gcm-motif (glial cell missing motif). The encoded protein is a homolog of the Drosophila glial cells missing gene (gcm). This protein binds to the GCM-motif (A/G)CCCGCAT, a novel sequence among known targets of DNA-binding proteins. The N-terminal DNA-binding domain confers the unique DNA-binding activity of this protein.

Specifications

Antigen GCM1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene GCM1
Gene Accession No. Q9NP62
Gene Alias chorion-specific transcription factor GCMa; Gcm 1; Gcm a; GCM motif protein 1; GCM1; Gcm1-rs1; Gcma; Gcm-rs2; glial cell deficient; glial cells missing homolog 1; glial cells missing homolog 1 (Drosophila); glial cells missing transcription factor 1; glide; hGCMa; mGCM1; mGCMa
Gene Symbols GCM1
Host Species Rabbit
Immunogen Recombinant full length Human GCM1.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8521
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less