missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GCLM Recombinant Rabbit Monoclonal Antibody (23GB5535)
GREENER_CHOICE

Catalog No. MA553894
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA553894 100 μL
MA553895 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA553894 Supplier Invitrogen™ Supplier No. MA553894
Only null left

Rabbit Recombinant Monoclonal Antibody

GCLM Recombinant Monoclonal Antibody for Flow, ICC/IF, Western Blot

Gamma-glutamylcysteine synthetase (gamma-GCS) is the rate limiting enzyme for glutathione (L-gamma-glutamyl-L-cysteinylglycine, GSH) synthesis. GSH is ubiquitous in mammalian cells as a vital intra- and extracellular protective antioxidant. Gamma-GCS is a heterodimer of a heavy catalytic subunit and a light regulatory subunit that is responsive to inflammation, phenolic ntioxidants, heat shock, oxidants and cytokines.

Specifications

Antigen GCLM
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 23GB5535
Concentration 0.18 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Gclm
Gene Accession No. O09172, P48507, P48508
Gene Alias AI649393; gamma glutamylcysteine synthetase; gamma-ECS regulatory subunit; Gamma-glutamylcysteine synthetase regulatory subunit; Gclm; Gcmc; GCS light chain; Glclr; glutamate cysteine ligase (gamma-glutamylcysteine synthetase), regulatory; glutamate cysteine ligase, modifier subunit; glutamate-cysteine ligase (gamma-glutamylcysteine synthetase), regulatory (30.8kD); glutamate-cysteine ligase modifier subunit; glutamate--cysteine ligase modifier subunit; glutamate-cysteine ligase modifier subunit delta2 alternative splicing; glutamate-cysteine ligase regulatory protein; Glutamate--cysteine ligase regulatory subunit; glutamate-cysteine ligase, modifier subunit; GSC light chain; null
Gene Symbols Gclm
Host Species Rabbit
Immunogen A synthesized peptide derived from human GCLM (65-100AA).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14630, 2730, 29739
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less