missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GCKR Monoclonal Antibody (4#)

Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA566122
Description
GCKR Monoclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA
This gene encodes a protein belonging to the GCKR subfamily of the SIS (Sugar ISomerase) family of proteins. The gene product is a regulatory protein that inhibits glucokinase in liver and pancreatic islet cells by binding non-covalently to form an inactive complex with the enzyme. This gene is considered a susceptibility gene candidate for a form of maturity-onset diabetes of the young (MODY).Chemical Identifiers
| GCKR | |
| 1 mg/mL | |
| ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Mouse | |
| RUO | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| Q07071, Q14397, Q91X44 | |
| 231103, 25658, 2646 | |
| Recombinant Glucokinase Regulatory Protein (GKRP) (Val90-Val286). | |
| Antibody |
| 4# | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Monoclonal | |
| Liquid | |
| IgG1 κ | |
| Human, Mouse, Rat | |
| Gckr | |
| FGQTL5; Gckr; GKRP; GLRE; glucokinase (hexokinase 4) regulator; glucokinase regulator; Glucokinase regulatory protein; hexokinase 4 | |
| Gckr | |
| Primary | |
| Protein A/G |
Specifications
| GCKR | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| Q07071, Q14397, Q91X44 | |
| Gckr | |
| Recombinant Glucokinase Regulatory Protein (GKRP) (Val90-Val286). | |
| 200 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG1 κ |
| ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 4# | |
| Unconjugated | |
| Gckr | |
| FGQTL5; Gckr; GKRP; GLRE; glucokinase (hexokinase 4) regulator; glucokinase regulator; Glucokinase regulatory protein; hexokinase 4 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 231103, 25658, 2646 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |