missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GCDH, Mouse anti-Human, Polyclonal Antibody, Abnova™
Mouse Polyclonal Antibody
Supplier: Abnova Corporation H00002639B01P
Description
Sequence: MALRGVSVRLLSRGPGLHVLRTWVSSAAQTEKGGRTQSQLAKSSRPEFDWQDPLVLEEQLTTDEILIRDTFRTYCQERLMPRILLANRNEVFHREIISEMGELGVLGPTIKGYGCAGVSSVAYGLLARELERVDSGYRSAMSVQSSLVMHPIYAYGSEEQRQKYLPQLAKGELLGCFGLTEPNSGSDPSSMETRAHYNSSNKSYTLNGTKTWITNSPMADLFVVWARCEDGCIRGFLLEKGMRGLSAPRIQGKFSLRASATGMIIMDGVEVPEENVLPGASSLGGPFGCLNNARYGIAWGVLGASEFCLHTARQYALDRMQFGVPLARNQLIQKKLADMLTEITLGLHACLQLGRLKDQDKAAPEMVSLLKRNNCGKALDIARQARDMLGGNGISDEYHVIRHAMNLEAVNTYEGTHDIHALILGRAITGIQAFTASKSpecifications
| GCDH | |
| Polyclonal | |
| In 1x PBS, pH 7.4 | |
| NM_000159.2 | |
| GCDH | |
| GCDH (NP_000150.1, 1 a.a. ∼ 438 a.a) full-length human protein. | |
| RUO | |
| 2639 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Purified |
| Immunofluorescence, Western Blot | |
| Unconjugated | |
| glutaryl-Coenzyme A dehydrogenase | |
| ACAD5/GCD | |
| Mouse | |
| 50 μg | |
| Primary | |
| Human | |
| Antibody |