missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GBP7 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5149955
Description
GBP7 Polyclonal Antibody for Western Blot, IHC, ICC/IF
Guanylate-binding proteins, such as GBP7, are induced by interferon and hydrolyze GTP to both GDP and GMP.Specifications
| GBP7 | |
| Polyclonal | |
| Unconjugated | |
| GBP7 | |
| 9830147J24Rik; GBP4L; Gbp7; GBP-7; GTP-binding protein 7; guanine nucleotide-binding protein 7; guanylate binding protein 7; Guanylate-binding protein 4-like; Guanylate-binding protein 7 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 388646 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry, Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| Q8N8V2 | |
| GBP7 | |
| Recombinant Guanylate Binding Protein 7 (GBP7) (Thr35-Lys277). | |
| 200 μL | |
| Primary | |
| Human, Rat | |
| Antibody |