Learn More
Thermo Scientific™ pTK-Gaussia-Dura Luc Vector for Luciferase Assays
Catalog No. PI16192
Change view
Click to view available options
Quantity:
10 μg
Catalog No. Quantity
PI16192 10 μg
1 options
Catalog No. PI16192 Supplier Thermo Scientific™ Supplier No. 16192
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Drive mutated Gaussia luciferase expression with vectors containing a multiple cloning site (MCS) or control promoter (CMV or TK) for extended signal output.
Thermo Scientific™ pMCSGaussia-Dura Luc Vector is a multiple cloning site plasmid designed to accept a promoter sequence for glow-analysis of gene regulation using the naturally secreting Gaussia luciferase reporter.
The Gaussia Dura Luc Vectors contains a mutant form of the Gaussia luciferase gene that confers better bioluminescent signal stability than native luciferase. Gaussia-Dura luciferase (approx. 20kDa) is a secreted protein, which enables measurement of the reporter activity in media (for real-time assays) and in cell lysates. The pMCS vector contains a multiple cloning site for cloning a promoter to study its regulatory potential. The pCMV and pTK vectors have the luciferase gene under the CMV (Cytomegalovirus) promoter and Herpes Simplex Virus (HSV) thymidine kinase (TK) promoter, respectively. These constitutive expression vectors can be used as normalization controls to account for experimental variation in combination with other reporters.
Highlights:
  • Naturally-secreting Gaussia-Dura luciferase gene, optimized for high expression and glow-stability in mammalian systems
  • Multiple cloning site (MCS) provides versatility for transfer of regulatory elements from one plasmid to another
  • Transcription termination site (Ter), Lac operator (Lac O1), and transcriptional pause site (TPS) used to minimize background by reducing transcriptional read-through
  • Both puromycin (Pur) and ampicillin (Amp) markers for drug selection in mammalian and bacterial cells, respectively
  • High-copy pUC bacterial DNA replication origin
  • Two control vectors available with strong (CMV) or weak (TK) constitutive promoters for co-transfection and normalization
Specifications
Description pTK-Gaussia-Dura Luc Vector
Type Luc Vector
Cloning Method Restriction Enzyme/MCS
Delivery Type Transfection
Quantity 10 μg
Content And Storage Store in original container protected from direct sunlight in a dry, cool and well-ventilated area, between the following temperatures: -70 to -20°C.
Reporter Gene Gaussia Dura Luciferase
Selection Agent (Eukaryotic) Puromycin
For Use With (Application) Reporter Assays
Promoter TK
What options do you offer for luciferase vectors?
There are three vectors available for each expressed luciferase; pMCS, pCMV, and pTK. We recommend using pMCS to measure the activity of the reporter, and pCMV, and pTK as positive control vectors. For high level constitutive expression of Luciferase, we recommend using the CMV (Cytomegalovirus) promoter vector and for low level constitutive expression of luciferase, we recommend using the pTK (thymidine kinase) promoter vector.
What is the stability of the secreted luciferase?
Gaussia, Gaussia-Dura, and Cypridina Luciferase are stable in the culture media for greater than 24 hours. Generally, after transfection of CMV driven constructs, we see ever increasing secreted signal between 24 and 72 hours.
How quickly is luciferase secreted?
The earliest detectable signal will depend on the strength of the promoter. In general, with a strong promoter (such as CMV), significant signal over background can be seen in as little as 20 minutes. However, with a weaker inducible promoter, significant signal over background may take 1-2 hours after induction.
Do you still offer quantitative IP (qIP) Luciferase assay kits?
Our qIP Luciferase assay kits have been discontinued but we do carry epitope-tagged (HA or c-Myc) and Tluc (TurboLuc luciferase)-tagged mammalian expression vectors, and qIP protein interaction assay reagents for these assay kits.
For Research Use Only. Not for use in diagnostic procedures.