missing translation for 'onlineSavingsMsg'
Learn More

Gatifloxacin, 98%, Thermo Scientific Chemicals

Catalog No. AC460230010
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC460230010 1 g
AC460230050 5 g
AC460230250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC460230010 Supplier Thermo Scientific Chemicals Supplier No. 460230010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 112811-59-3
Molecular Formula C19H22FN3O4
Molecular Weight (g/mol) 375.40
MDL Number MFCD00895399
InChI Key XUBOMFCQGDBHNK-UHFFFAOYNA-N
Synonym gatifloxacin, tequin, zymar, gatiflo, gatifloxacine, gatiquin, gatispan, gatilox, zymaxid, gaity
PubChem CID 5379
ChEBI CHEBI:5280
IUPAC Name 1-cyclopropyl-6-fluoro-8-methoxy-7-(3-methylpiperazin-1-yl)-4-oxoquinoline-3-carboxylic acid
SMILES COC1=C2N(C=C(C(O)=O)C(=O)C2=CC(F)=C1N1CCNC(C)C1)C1CC1

Specifications

Melting Point 185.0°C to 187.0°C
Loss on Drying 4% to 8%
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 375.39
Percent Purity 98%
Chemical Name or Material Gatifloxacin
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing.
IF ON SKIN: Wash with plenty of soap and water.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove to fresh air and keep at rest in a position

RUO – Research Use Only