missing translation for 'onlineSavingsMsg'
Learn More

Accuform Signs Gate Valve Lockouts

Available in five sizes these valve covers ensure no one will be able to operate valve handles

$99.94 - $99.94

Specifications

Product Type Gate Valve Lockout
Color Red
Material Polypropylene
For Use With (Application) To enclose and fully surround the valve handle
Temperature Range (English) −50° to +360°F
View More Specs

Products 1
Catalog Number Mfr. No. Diameter (English) Price Quantity  
Catalog Number Mfr. No. Diameter (English) Price Quantity  
19-804-546
View Documents
Accuform Signs
KDD474
10 to 13 in.
Each for $99.94
Only null left
 
Description

Description

Specifications

Specifications

Gate Valve Lockout
Polypropylene
−50° to +360°F
Red
To enclose and fully surround the valve handle
−46° to +182°C
Videos
SDS
Documents

Documents