missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GATAD2A Recombinant Rabbit Monoclonal Antibody (23GB5685)

Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA553976
Description
GATAD2A Recombinant Monoclonal Antibody for Flow, ICC/IF, Western Blot
GATAD2A is a gene encoding a protein that functions as a subunit of the nucleosome remodeling and deacetylase (NuRD) complex, which plays a crucial role in chromatin remodeling and gene expression regulation. Located on the human chromosome, GATAD2A contains proline-rich PPPL theta motifs that facilitate interactions with MYND domains in proteins such as ZMYND8, distinguishing it from its paralog GATAD2B. This gene is important for various cellular processes, particularly during neural development, where it participates in modulating chromatin structure through histone deacetylation and ATP-dependent remodelings, like those mediated by NuRD. Moreover, GATAD2A has been implicated in certain neurodevelopmental disorders, suggesting its distinct functional roles in cellular contexts. Additionally, research has identified circular RNA variants of GATAD2A, which have been shown to influence viral replication, indicating its broader impact on cellular function beyond chromatin modification.Chemical Identifiers
| GATAD2A | |
| 1.5 mg/mL | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q86YP4, Q8CHY6 | |
| 234366, 54815 | |
| A synthesized peptide derived from human p66 alpha (175-236AA). | |
| Antibody |
| 23GB5685 | |
| -20°C | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Human, Mouse | |
| GATAD2A | |
| 1110066C11Rik; BC031407; C80248; GATA zinc finger domain containing 2A; GATA zinc finger domain-containing protein 2A; GATAD2A; Hp66alpha; MGC29315; MGC36975; p66 alpha; p66a; p66alpha; RGD1310596; transcriptional repressor p66 alpha; Transcriptional repressor p66-alpha | |
| GATAD2A | |
| Primary | |
| Affinity chromatography |
Specifications
| GATAD2A | |
| Recombinant Monoclonal | |
| 1.5 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q86YP4, Q8CHY6 | |
| GATAD2A | |
| A synthesized peptide derived from human p66 alpha (175-236AA). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB5685 | |
| Unconjugated | |
| GATAD2A | |
| 1110066C11Rik; BC031407; C80248; GATA zinc finger domain containing 2A; GATA zinc finger domain-containing protein 2A; GATAD2A; Hp66alpha; MGC29315; MGC36975; p66 alpha; p66a; p66alpha; RGD1310596; transcriptional repressor p66 alpha; Transcriptional repressor p66-alpha | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 234366, 54815 | |
| -20°C | |
| Liquid |