missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GAS8 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559509
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559509 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559509 Supplier Invitrogen™ Supplier No. PA559509
Only null left

Rabbit Polyclonal Antibody

Immunogen sequence: VEKKEVQFNE VLAASNLDPA ALTLVSRKLE DVLESKNSTI KDLQYELAQV CKAHNDLLRT YEAKLLAFGI PLDNVGFKPL ETAVIGQTLG QG Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 96%.

This gene includes 11 exons spanning 25 kb and maps to a region of chromosome 16 that is sometimes deleted in breast and prostrate cancer. The second intron contains an apparently intronless gene, C16orf3, that is transcribed in the opposite orientation. This gene is a putative tumor suppressor gene.

Specifications

Antigen GAS8
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene GAS8
Gene Accession No. O95995
Gene Alias beta growth arrest specific protein 11; CILD33; DRC4; Dynein regulatory complex subunit 4; Gas11; GAS-11; Gas8; GAS-8; growth arrest specific 11; growth arrest specific 8; growth arrest specific protein 11; growth arrest-specific protein 11; Growth arrest-specific protein 8; MGC138326
Gene Symbols GAS8
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human GAS8. Recombinant protein control fragment (Product #RP-98688).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2622
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less