missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GAS2, Mouse anti-Human, Polyclonal Antibody, Abnova™
Mouse Polyclonal Antibody
Supplier: Abnova Corporation H00002620B01P
Description
Sequence: MCTALSPKVRSGPGLSDMHQYSQWLASRHEANLLPMKEDLALWLTNLLGKEITAETFMEKLDNGALLCQLAETMQEKFKESMDANKPTKNLPLKKIPCKTSAPSGSFFARDNTANFLSWCRDLGVDETCLFESEGLVLHKQPREVCLCLLELGRIAARYGVEPPGLIKLEKEIEQEETLSAPSPSPSPSSKSSGKKSTGNLLDDAVKRISEDPPCKCPNKFCVERLSQGRYRVGEKILFIRMLHNKHVMVRVGGGWETFAGYLLKHDPCRMLQISRVDGKTSPIQSKSPTLKDMNPDNYLVVSASYKAKKEIKSpecifications
| GAS2 | |
| Polyclonal | |
| In 1x PBS, pH 7.4 | |
| NM_005256 | |
| GAS2 | |
| GAS2 (NP_005247.1, 1 a.a. ∼ 313 a.a) full-length human protein. | |
| RUO | |
| 2620 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Purified |
| Western Blot | |
| Unconjugated | |
| growth arrest-specific 2 | |
| MGC32610 | |
| Mouse | |
| 50 μg | |
| Primary | |
| Human | |
| Antibody |