missing translation for 'onlineSavingsMsg'
Learn More

Justrite™ Gas Cylinder Stand

Justrite™ Gas Cylinder Stand is designed for the safe storage of industrial and commercial use compressed gas cylinders. Help prevent dangerous tipping situations

Supplier:  Justrite™ 35290

 View more versions of this product

Catalog No. 19085828


Only null left

Description

Description

  • Fully welded construction using steel or stainless steel provides structural integrity and longevity
  • Exclusive bumper guard edging with steel reinforced vinyl protects your cylinders from damage while providing extra grip
  • Accepts cylinders from 4 to 12in (102 to 305mm) in diameter
  • Pre-drilled holes in base for floor mounting with application specific fasteners (not included)
  • Heavy-duty polypropylene support straps, 1.5 x 54in (38mm x 1372mm) and rated at 1138lb (516kg), feature steel cinch buckle and hold individual cylinders securely against integral bracket
  • For an improved level of safety for any installation, all brackets include openings to retrofit secondary support chains, one set for each cylinder strap location

Compliance

OSHA, CGA P1, NFPA55

Warranty

1 YEAR

Specifications

Chemical Identifiers

Gas Cylinder Stand
Gas cylinders
762 mm

Specifications

Gas Cylinder Stand
Stainless Steel
61 x 32 x 28in.
762 mm
19 lb.
2 Cylinders
Silver
Gas cylinders
267 mm
705 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions