missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Justrite™ Gas Cylinder Barricade Rack
Justrite™ Gas Cylinder Barricade Rack secures cylinders up to 12-in (305-mm) in diameter
Supplier: Justrite™ 35140
Description
- Meets USGS high-hazard seismic zone requirements when properly mounted
- Recommended for indoor or outdoor bulk cylinder storage
- Surround cylinders with 2-in (51-mm) square steel structural tubing
- Continuously seam welded at all junction points
- Sealed with exterior grade powder-coat paint for long lasting strength, durability and dependability
- Tops ground smooth and capped with high-density polyethylene (HDPE) caps to prevent corrosion and injuries
- Features two levels of welded steel chains for each cylinder and clasps to connect the chains to the rack, ensuring a snug fit
- Pre-drilled 0.531-in (14-mm) diameter floor mounting holes included on lower tube structure
- For seismic installations, add Model 35350 S4-rated anchor bolts (not included)
Compliance
OSHA, CGA P1, NFPA55
Warranty
1 YEAR
Chemical Identifiers
| Steel | |
| Black | |
| Gas cylinder | |
| 762 mm |
Specifications
| 12 Cylinders: 3 Wide by 4 Deep | |
| Black | |
| 762 mm | |
| 114 lb. |
| Steel | |
| Gas cylinder | |
| 1118 mm | |
| 1118 mm |