missing translation for 'onlineSavingsMsg'
Learn More

Justrite™ Gas Cylinder Barricade Rack

Catalog No. 19085776
Change view
Click to view available options
Holds:
1 Cylinder: 1 Wide by 1 Deep
10 Cylinders: 5 Wide by 2 Deep
12 Cylinders: 3 Wide by 4 Deep
12 Cylinders: 4 Wide by 3 Deep
15 Cylinders: 5 Wide by 3 Deep
16 Cylinders: 4 Wide by 4 Deep
2 Cylinders: 1 Wide by 2 Deep
2 Cylinders: 2 Wide by 1 Deep
20 Cylinders: 5 Wide by 4 Deep
3 Cylinders: 1 Wide by 3 Deep
3 Cylinders: 3 Wide by 1 Deep
4 Cylinders: 1 Wide by 4 Deep
4 Cylinders: 2 Wide by 2 Deep
4 Cylinders: 4 Wide by 1 Deep
5 Cylinders: 5 Wide by 1 Deep
6 Cylinders: 2 Wide by 3 Deep
6 Cylinders: 3 Wide by 2 Deep
8 Cylinders: 2 Wide by 4 Deep
8 Cylinders: 4 Wide by 2 Deep
9 Cylinders: 3 Wide by 3 Deep
20 product options available for selection
Product selection table with 20 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Holds
19085776 1 Cylinder: 1 Wide by 1 Deep
19085777 2 Cylinders: 1 Wide by 2 Deep
19085778 3 Cylinders: 1 Wide by 3 Deep
19085779 4 Cylinders: 1 Wide by 4 Deep
19085780 2 Cylinders: 2 Wide by 1 Deep
19085781 4 Cylinders: 2 Wide by 2 Deep
19085782 6 Cylinders: 2 Wide by 3 Deep
19085783 8 Cylinders: 2 Wide by 4 Deep
19085784 3 Cylinders: 3 Wide by 1 Deep
19085785 6 Cylinders: 3 Wide by 2 Deep
19085787 9 Cylinders: 3 Wide by 3 Deep
19085789 12 Cylinders: 3 Wide by 4 Deep
19085790 4 Cylinders: 4 Wide by 1 Deep
19085791 8 Cylinders: 4 Wide by 2 Deep
19085792 12 Cylinders: 4 Wide by 3 Deep
19085793 16 Cylinders: 4 Wide by 4 Deep
19085794 5 Cylinders: 5 Wide by 1 Deep
19085795 10 Cylinders: 5 Wide by 2 Deep
19085796 15 Cylinders: 5 Wide by 3 Deep
19085797 20 Cylinders: 5 Wide by 4 Deep
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 20 options available.
20 options
Catalog No. 19085776 Supplier Justrite™ Supplier No. 35080

Justrite™ Gas Cylinder Barricade Rack secures cylinders up to 12-in (305-mm) in diameter

  • Meets USGS high-hazard seismic zone requirements when properly mounted
  • Recommended for indoor or outdoor bulk cylinder storage
  • Surround cylinders with 2-in (51-mm) square steel structural tubing
  • Continuously seam welded at all junction points
  • Sealed with exterior grade powder-coat paint for long lasting strength, durability and dependability
  • Tops ground smooth and capped with high-density polyethylene (HDPE) caps to prevent corrosion and injuries
  • Features two levels of welded steel chains for each cylinder and clasps to connect the chains to the rack, ensuring a snug fit
  • Pre-drilled 0.531-in (14-mm) diameter floor mounting holes included on lower tube structure
  • For seismic installations, add Model 35350 S4-rated anchor bolts (not included)

Compliance

OSHA, CGA P1, NFPA55

Warranty

1 YEAR

Specifications

Holds 1 Cylinder: 1 Wide by 1 Deep
Material Steel
Color Black
For Use With (Equipment) Gas cylinder
Height (Metric) 762 mm
Length (Metric) 406 mm
Weight (English) 26 lb.
Width (Metric) 406 mm