missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GARS Recombinant Rabbit Monoclonal Antibody (23GB5515)
GREENER_CHOICE

Catalog No. MA553892
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA553892 100 μL
MA553893 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA553892 Supplier Invitrogen™ Supplier No. MA553892
Only null left

Rabbit Recombinant Monoclonal Antibody

GARS Recombinant Monoclonal Antibody for Flow, ICC/IF, Western Blot

GARS is a glycyl-tRNA synthetase, one of the aminoacyl-tRNA synthetases that charge tRNAs with their cognate amino acids. This protein is an (alpha)2 dimer which belongs to the class II family of tRNA synthetases. The protein has been shown to be a target of autoantibodies in the human autoimmune diseases, polymyositis or dermatomyositis.

Specifications

Antigen GARS
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 23GB5515
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene GARS1
Gene Accession No. P41250, Q5I0G4, Q9CZD3
Gene Alias 6.1.1.14; AIR synthase; AIRS; Ap4A synthetase; AP-4-A synthetase; Charcot-Marie-Tooth neuropathy 2D; Charcot-Marie-Tooth neuropathy, neuronal type, D; CMT2D; Diadenosine tetraphosphate synthetase; DSMAV; EC 2.1.2.29; GAR transformylase; GARS; gars1; GART; GARTF; Gena201; GENA202; glycine tRNA ligase; glycine--tRNA ligase; glycyl-tran synthetase; glycyl-tRNA synthetase; glycyl-tRNA synthetase 1; glycyl-tRNA synthetase-like protein; glyRS; HMN5; metronome; MGC47764; mrn; Nmf249; PAIS; PGFT; PRGS; RGD1559871; Sgrp23; si:dkey-276i5.1; SMAD1; storage granule protein 23; Unknown (protein for IMAGE:7254992); wu:fb02f03
Gene Symbols GARS, GARS1
Host Species Rabbit
Immunogen A synthesized peptide derived from human GARS (701-739AA).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2617, 297113, 353172
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less