missing translation for 'onlineSavingsMsg'
Learn More

(+)-Garcinia acid, ≥95.0% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111018488
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111018488 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111018488 Supplier Merck Emd Millipore Supplier No. 4429210MG
Featured Industries: Food and Beverages

Specifications

Packaging Bottomless Glass Bottle with Inserted Fused Cone
CAS 27750-13-6
Physical Form Neat
Linear Formula C6H6O7
MDL Number MFCD06656311
Synonym (-)-Hydroxycitric acid lactone; (2 S,3 S)-3-Hydroxy-5-oxotetrahydrofuran-2,3-dicarboxylic acid; Garcinia lactone
Type Analytical Standard
Quantity 10 mg
Formula Weight 190.11
Percent Purity ≥95.0% (HPLC)
Grade Analytical Standard
Show More Show Less