missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GAP43 Monoclonal Antibody (K29024_1C6), Invitrogen™
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA555647
Description
Sequence of this protein is as follows: VEKNDDDQKI EQDGIKPEDK AHKAATKIQA SFRGHITRKK LKGEKKDDVQ AAEAEANKKD EAPVADGVEK KGEGTTTAEA APATGSKPDE PGKAGETPSE
GAP43 (Neuromodulin) is associated with nerve growth. GAP43 is expressed by developing and regenerating neurons, and to a lesser extent reactive glial cells. It is a major component of the motile 'growth cones' that form the tips of elongating axons. GAP43 also plays a role in axonal and dendritic filopodia induction.Specifications
| GAP43 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| P07936, P17677 | |
| GAP43 | |
| Recombinant human GAP43 (immunogen range is 11-110). | |
| 50 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG2b |
| Immunoprecipitation, Western Blot | |
| K29024_1C6 | |
| Unconjugated | |
| GAP43 | |
| Axonal membrane protein GAP-43; B-50; Basp2; brain abundant, membrane attached signal protein 2; calmodulin-binding protein P-57; Gap43; GAP-43; growth accentuating protein 43; growth associated protein 43; growth-associated protein 43; MGC156666; nerve growth-related peptide GAP43; neural phosphoprotein B-50; Neuromodulin; neuron growth-associated protein 43; neuron protein F1; PP46; Protein F1 | |
| Mouse | |
| Protein A | |
| RUO | |
| 2596, 29423 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |