missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Ganglioside GM2 Chimeric Recombinant Rabbit Monoclonal Antibody (KM966)

Description
Specificity: This antibody recognizes and binds ganglioside GM2.
Ganglioside GM2 is a glycosphingolipid found in the plasma membrane of cells, particularly in the nervous system. It is composed of a ceramide lipid tail and a complex carbohydrate head group that contains sialic acid residues. The function of Ganglioside GM2 is not fully understood, but it is believed to play a role in cell signaling and communication, as well as in the formation and maintenance of the myelin sheath that surrounds nerve cells. Mutations in the genes responsible for the synthesis or degradation of Ganglioside GM2 can result in various neurological disorders, including Tay-Sachs disease and Sandhoff disease.
Specifications
Specifications
| Antigen | Ganglioside GM2 Chimeric |
| Applications | ELISA, Flow Cytometry, Functional Assay, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Clone | KM966 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% ProClin 300 |
| Host Species | Rabbit |
| Immunogen | This chimeric antibody was generated by cloning the variable heavy and light chain sequences of murine parent antibody KM696 (IgM) and expressing it as a mouse/human IgG1. The parent antibody was raised against human GM2. |
| Purification Method | Protein A |
| Show More |