missing translation for 'onlineSavingsMsg'
Learn More

Ganciclovir Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111012247
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111012247 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111012247 Supplier Merck Emd Millipore Supplier No. PHR15931G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.Ganciclovir is a nucleoside analog which is utilized extensively for the remedy of cytomegalovirus infections. It is also found to be efficient in the treatment of cytomegalovirus in immunocompromised patients, particularly against acquired immunodeficiency syndrome (AIDS) and congenital immunodeficiency. Featured Industries: Pharmaceutical (small molecule)

Specifications

CAS 82410-32-0
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Linear Formula C9H13N5O4
MDL Number MFCD00870588
Synonym Ganciclovir; 2-Amino-1,9-dihydro-9-[[2-hydroxy-1-(hydroxymethyl)ethoxy]methyl]-6 H-purin-6-one; 9-(1,3-Dihydroxy-2-propoxymethyl)guanine; DHPG
Type Certified Reference Material
Quantity 1 g
Formula Weight 255.23
Grade Certified Reference Material